Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4502926

Sigma-Aldrich

Anti-TBP antibody produced in rabbit

affinity isolated antibody

Synonym(s):

TATA binding factor, TATA sequence-binding protein, TATA-box binding protein, TATA-box factor, TFIID

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 37 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TBP(6908)

General description

Anti-TBP antibody detects endogenous levels of total TBP protein.
The TBP (TATA-binding protein) gene is mapped to human chromosome 6q27.

Immunogen

The antiserum was produced against synthesized peptide derived from human TBP.

Immunogen Range: 151-200

Biochem/physiol Actions

The TBP (TATA-binding protein) gene encodes a subunit of TFIIIB (transcription factor IIIB) complex and recognizes the TATA box in the DNA. It thus, plays an important role in RNA polymerase II transcription machinery. TBP is known to induce stress granule formation in hepatitis B viral infection.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doubly Spliced RNA of Hepatitis B Virus Suppresses Viral Transcription via TATA-Binding Protein and Induces Stress Granule Assembly.
Tsai KN
Journal of Virology, 89(22), 11406-11419 (2015)
The Pathogenic Role of Low Range Repeats in SCA17.
Shin JH
PLoS ONE, 10(8), 1-10 (2015)
Taspase1 processing alters TFIIA cofactor properties in the regulation of TFIID.
Malecova B
Transcription, 6(2), 21-32 (2015)
Molecular mechanisms of Bdp1 in TFIIIB assembly and RNA polymerase III transcription initiation.
Gouge J
Nature Communications, 8(1):130, 1-11 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service