Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-1531

Sigma-Aldrich

Anti-TATA-box-binding protein Antibody, clone SL30-3-563

clone SL30-3-563, from mouse

Synonym(s):

TATA box binding protein, Transcription initiation factor TFIID TBP subunit, TATA-binding factor, TATA sequence-binding protein, TATA-box binding protein N-terminal domain, TATA-box factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SL30-3-563, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TBP(6908)

General description

The TATA-box-binding protein (TBP) is a critical, eukaryotic transcription factor. TBP a part of the RNA polymerase II eukaryotic transcription complex and is required for the transcriptional initiation of RNA polymerases. TBP is considered to be a limiting factor for certain RNA promoters. Expression is ubiquitous, however the highest expression has been observed in testis and ovary. Mutations in TBP result in spinocerebellar ataxia type 17(SCA17), a cerebellar disorder classified as autosomal dominant cerebellar ataxia and characterized by atrophy to the cerebellar and cerebral regions, extrapyramidal signs, and dementia.

Specificity

This antibody recognizes TATA-box-binding protein at the N-terminus.

Immunogen

Epitope: N-terminus
GST-tagged recombinant protein corresponding to human TATA-box-binding protein at the N-terminus.

Application

Chromatin Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in U2OS cells. (Denisso, S., et al. (2007). The EMBO Journal. 26:944–954.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-TATA-box-binding protein Antibody, clone SL30-3-563 (mouse monoclonal antibody) validated in WB to detect TATA-box-binding protein also known as TATA box binding protein, Transcription initiation factor TFIID TBP subunit.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected TATA-box-binding protein on 10 µg of HeLa cell lysate.

Target description

~38 kDa observed.

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Wille et al.
Behavioural brain research, 311, 141-146 (2016-05-22)
Enhanced anxiety is a salient feature of a number of psychiatric disorders including anxiety disorders, trauma-related disorders and depression. Although aberrant expression of various genes has been detected in patients suffering from persistent high anxiety as well as in high
Alexandra Wille et al.
Frontiers in behavioral neuroscience, 9, 313-313 (2015-12-05)
Successful attenuation of fearful memories is a cognitive process requiring initiation of highly coordinated transcription programs. Chromatin-modulating mechanisms such as DNA methylation and histone modifications, including acetylation, are key regulators of these processes. However, knowledge concerning the role of ATP-dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service