Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4502713

Sigma-Aldrich

Anti-SODIUM CHANNEL-PAN antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CIN5, HH1, SCN5A, Sodium channel protein, cardiac muscle α-subunit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 227 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCN2A(6323)

General description

Anti-SODIUM CHANNEL-PAN Antibody detects endogenous levels of total SODIUM CHANNEL-PAN protein.
The SCN5A (sodium channel protein type 5 subunit α) gene is mapped to human chromosome 3p22.2. The encoded protein is known to be localized to cardiac cells.

Immunogen

The antiserum was produced against synthesized peptide derived from human Sodium Channel.

Immunogen Range: 1466-1515

Biochem/physiol Actions

SCN5A (sodium channel protein type 5 subunit α) is associated with the regulation of myocardial excitability as it conducts cardiac depolarization via inward Na+ current. Mutation in SCN5A causes retarded cardiac repolarization resulting in congenital long QT syndrome and also causes Brugada syndrome due to loss of gene function.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrophysiological characterization of a large set of novel variants in the SCN5A-gene: identification of novel LQTS3 and BrS mutations.
Ortiz-Bonnin B
Pfluegers Archiv, 468(8), 1375-1387 (2016)
C-terminal phosphorylation of NaV1.5 impairs FGF13-dependent regulation of channel inactivation.
Burel S
The Journal of Biological Chemistry, 292(42), 17431-17448 (2017)
Sirtuin 1 regulates cardiac electrical activity by deacetylating the cardiac sodium channel.
Vikram A
Nature Medicine, 23(3), 361-367 (2017)
A common genetic variant within SCN10A modulates cardiac SCN5A expression.
van den Boogaard M
The Journal of Clinical Investigation, 124(4), 1844-1852 (2014)
Shehla U Hridi et al.
Molecular and cellular neurosciences, 95, 71-78 (2019-02-10)
Interleukin 16 (IL-16) is a cytokine that is primarily associated with CD4+ T cell function, but also exists as a multi-domain PDZ protein expressed within cerebellar and hippocampal neurons. We have previously shown that lymphocyte-derived IL-16 is neuroprotective against excitotoxicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service