Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4501720

Sigma-Aldrich

Anti-LAMA5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

α 5, laminin, laminin α 5, laminin α-5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 399 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMA5(3911)

General description

Anti-LAMA5 Antibody detects endogenous levels of total LAMA5 protein. Laminin alpha5 (LAMA5) codes for the laminin α5 chain. It is mapped to human chromosome 20q13.2- q13.3. In human skin, LAMA5 is produced and liberated by dermal pericytes.

Immunogen

The antiserum was produced against synthesized peptide derived from human LAMA5.

Immunogen Range: 2381-2430

Application

Anti-LAMA5 antibody has been used in immunohistochemistry, immunocytochemistry and immunofluorescent staining.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Furthermore, studies in mice showed that Lama5 is involved in organogenesis and placental function during pregnancy. LAMA5 may play a role in regulating inter-individual variability in metabolic phenotypes in children. It induces skin regeneration both in vitro and in vivo.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A role for pericytes as microenvironmental regulators of human skin tissue regeneration.
Paquet-Fifield S, et al.
The Journal of Clinical Investigation, 119(9), 2795-2806 (2009)
Common variants in the LAMA5 gene associate with fasting plasma glucose and serum triglyceride levels in a cohort of pre-and early pubertal children.
De Luca M, et al.
Journal of pediatric genetics, 1(4), 229-234 (2012)
Glícia Maria de Almeida et al.
Scientific reports, 5, 17860-17860 (2015-12-15)
Extracellular matrix (ECM) is known to play several important roles in vascular development, although the molecular mechanisms behind these remain largely unknown. RECK, a tumor suppressor downregulated in a wide variety of cancers, encodes a membrane-anchored matrix-metalloproteinase-regulator. Mice lacking functional
Jyoti Gautam et al.
Scientific reports, 6, 36450-36450 (2016-11-04)
Laminin, a major component of the basement membrane, plays an important role in blood brain barrier regulation. At the neurovascular unit, brain endothelial cells, astrocytes, and pericytes synthesize and deposit different laminin isoforms into the basement membrane. It has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service