Skip to Content
MilliporeSigma
All Photos(18)

Key Documents

ZRB1023

Sigma-Aldrich

Anti-Laminin α2 Antibody, clone 2I22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Laminin M chain, Laminin-12 subunit alpha, Laminin-2 subunit alpha, Laminin-4 subunit alpha, Merosin heavy chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2I22, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 343.91 kDa
observed mol wt ~350 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... LAMA2(3908)
mouse ... Lama2(16773)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2I22 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Laminin subunit alpha-2. It targets an epitope within 18 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal region of human Laminin 2 alpha.

Application

Anti-Laminin a2, clone 2I22 ZooMAb, Cat. No. ZRB1023, is a recombinant Rabbit monoclonal antibody that specifically targets Laminin 2 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blo
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Laminin 2 in human heart tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Laminin 2a peptide with a KD of 1.0 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Laminin 2 in AC16 and C2C12 cells.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected Laminin α2 in human heart tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Laminin a2 (UniProt: P24043; also known as Laminin M chain, Laminin-12 subunit alpha, Laminin-2 subunit alpha, Laminin-4 subunit alpha, Merosin heavy chain) is encoded by the LAMA2 (also known as LAMM) gene (Gene ID: 3908) in human. Laminins are a family of heterotrimeric extracellular glycoproteins consisting of alpha, beta, and gamma chains that are bound to each other by disulfide bonds into a cross-shaped molecule comprising one long and three short arms with globules at each end. Laminins mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Thus far five alpha, three beta, and three gamma subunits are known, which assemble into at least 15 different laminin molecules. Laminin a2 is a glycoprotein with multiple EGF-like domains and is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off. It is expressed in multiple tissues, including placenta, striated muscle, cardiac muscle, pancreas, lung, spleen, skin, bone, and some regions of the brain. Laminin a2 binds to cells via a high affinity receptor and mediates the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Mutations in LAMA2 gene have been linked Merosin-deficient congenital muscular dystrophy 1A (MDC1A) and limb-girdle muscular dystrophy that results in muscle weakness and atrophy. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service