Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4301471

Sigma-Aldrich

Anti-phospho-p70 S6 Kinase (pThr389/412) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KS6B1, P70-S6K, RPS6KB1, Ribosomal protein S6 kinase, polypeptide 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

70 kDa

species reactivity

human, mouse, rat

concentration

1.0 mg/mL

technique(s)

western blot: 1:500-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_001258971.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr389/pThr412)

Gene Information

human ... RPS6KB1(6198)

Specificity

The antibody detects endogenous level of p70 S6 Kinase only when phosphorylated at tyrosine 389/tyrosine 412.

Immunogen

Peptide sequence around phosphorylation site of threonine389/412(G-F-T(p)-Y-V) derived from Human p70 S6 Kinase .

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guanli Zhang et al.
Journal of biochemical and molecular toxicology, 37(1), e23237-e23237 (2022-10-05)
Pre-eclampsia (PE) is a major cause of hypertension in maternal and fetal. Atlastin-1 (ATL1), one regulator of endoplasmic reticulum (ER) morphology, participates in tubular ER formation and protein synthesis. The objective of this study is to investigate the role and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service