Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200252

Sigma-Aldrich

Anti-methyl-Histone H4 (Me-Lys20)antibody, Mouse monoclonal

clone 5E10-D8, purified from hybridoma cell culture

Synonym(s):

Anti-H4K20me1, MonoclonalAnti-HIST4H4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

5E10-D8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~14 kDa

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect immunofluorescence: 2.5-5.0 μg/mL using mouse 3T3 cells or rat NRK cells
western blot: 1.0-2.0 μg/mL using histones isolated from human HeLa cells.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

monomethylation (Lys20)

Gene Information

human ... H4-16(121504)

Immunogen

peptide containing methylated lysine 20 of human Histone H4, conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service