Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GBL246870

Grace Bio-Labs ProPlate® tray set

for spring clip modules

Synonym(s):

slide holder, slide module

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.15

sterility

non-sterile

manufacturer/tradename

Grace Bio-Labs 246870

external size

85.3 mm × 127.5 mm

color

none

Looking for similar products? Visit Product Comparison Guide

General description

holds up to 3 slides, includes cover, applicator, and 10 seal strips

Legal Information

ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service