Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200230

Sigma-Aldrich

Anti-DPP4/CD26 antibody, Mouse monoclonal

clone A6H, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-ADABP, Monoclonal Anti-ADCP2, Monoclonal Anti-DPPIV, Monoclonal Anti-TP103, Monoclonal Anti-dipeptidyl-peptidase 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

A6H, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~4.0 mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: 20-40 μg/mL using methanol/acetone fixed HT-29 cells

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DPP4(1803)

Related Categories

General description

Monoclonal Anti-DPP4/CD26 (mouse IgG1 isotype) is derived from the hybridoma A6H produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with human fetal homogenates. Dipeptidyl peptidase-4 (DPP4) belongs to the S9B protein family, which cleaves X-proline dipeptides from the N-terminus of polypeptides, such as chemokines, neuropeptides and peptide hormones. It is a type II transmembrane glycoprotein, expressed on the surface of many cell types.

Immunogen

human fetal homogenates.

Application

Monoclonal Anti-DPP4/CD26 antibody produced in mouse has been used in:
  • enzyme linked immuno sorbent assay (ELISA)
  • immunocytochemistry
  • immunohistochemistry
  • flow cytometry

Biochem/physiol Actions

Dipeptidyl peptidase-4 (DPP4) is involved in the regulation of the biological activity of hormones and chemokines, such as glucagon-like peptide-1 and glucose-dependent insulinotropic polypeptide. DPP4 inhibition maybe used to treat type 2 diabetes mellitus. It acts as a T cell activation molecule and regulates immunity and immune-mediated disorders.

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dipeptidyl peptidase-4 (CD26): knowing the function before inhibiting the enzyme
Matteucci E and Giampietro O
Current Medicinal Chemistry, 16(23), 2943-2951 (2009)
R L Vessella et al.
Cancer research, 45(12 Pt 1), 6131-6139 (1985-12-01)
Monoclonal antibodies (MABs) reactive with human renal cell carcinoma (RCC) were generated following immunization of mice with either RCC homogenates, RCC cell lines, or fetal kidney homogenates. The characteristics of four highly reactive immunoglobulin G1 MABs, designated UMVA-RCC-A6H, UMVA-RCC-A36, UMVA-RCC-C5H
Vlad Tocan et al.
The Journal of biological chemistry, 297(6), 101354-101354 (2021-11-01)
Hepatocytes differ from columnar epithelial cells by their multipolar organization, which follows the initial formation of central lumen-sharing clusters of polarized cells as observed during liver development and regeneration. The molecular mechanism for hepatocyte polarity establishment, however, has been comparatively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service