Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3501255

Sigma-Aldrich

Anti-Seasonal H1N1 Nucleocapsid Protein antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

Influenza A virus subtype H1N1

concentration

1 mg/mL

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Seasonal H1N1 Nucleocapsid Protein Antibody: Influenza A virus is a major public health threat, killing more than 30, 000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus (S-OIV) was identified in specimens obtained from patients in Mexico and the United States. The influenza A virus polymerase transcribes and replicates eight virion RNA (vRNA) segments, among which the nucleocapsid protein (NP), thought to control whether mRNA or cRNA is produced. The nucleoprotein (NP), which has multiple functions during the virus life cycle, possesses regions that are highly conserved among influenza A, B, and C viruses. It was recently found several NP mutations that affected the efficient incorporation of multiple viral-RNA (vRNA) segments into progeny virions even though a single vRNA segment was incorporated efficiently. This indicates that the respective conserved amino acids in NP may be critical for the assembly and/or incorporation of sets of eight vRNA segments.

Immunogen

NP antibody was raised against a synthetic peptide from the seasonal H1N1 NP protein.The immunogen is located within amino acids 350 - 400 of Seasonal H1N1 Nucleocapsid Protein.

Application

NP antibody can be used for the detection of the NP protein from the H1N1 strain of Seasonal Influenza A in ELISA.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service