Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA001465

Sigma-Aldrich

Anti-C7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Complement component C7 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

SAFLCGSSLKWSPEMKNARCVQKENPLTQAVPKCQRWEKLQNSRCVCKMPYECGPSLDVCAQDERSKRILPLTVCKMHVLHCQGRNYTLTGRDSCTLPASAEKACGACPLWGKCDAESSKCVCREASECEEEGFSICVEVNGKEQTMSEC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C7(730)

Related Categories

Immunogen

Complement component C7 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-C7 antibody produced in rabbit is suitable for global protein profiling to find new molecular biomarkers for common, multifactorial disorders.
Anti-C7 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

C7 (complement component 7) encodes an anchor protein that forms a component of the membrane attack complex (MAC). The complex forms pores in the plasma membrane of target cells and functions in the innate and adaptive immune response. Mutations in this gene can cause a deficiency or complete absence of C7 protein. This causes a rare defect in the complement classical pathway resulting in severe recurrent infections, mainly by Neisseria gonorrhoeae or Neisseria meningitidis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78246

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Sircar et al.
The American journal of pathology, 188(10), 2147-2154 (2018-09-27)
There is a temporal window from the time diabetes is diagnosed to the appearance of overt kidney disease during which time the disease progresses quietly without detection. Currently, there is no way to detect early diabetic nephropathy (EDN). Herein, we
Taco W Kuijpers et al.
Molecular immunology, 47(4), 671-677 (2009-11-26)
Meningococcal disease is caused by Neisseria meningitidis which is associated with high morbidity and mortality. Recurrences of meningococcal infection have been observed in patients with terminal complement component defects, because of the inefficient formation of the lytic membrane attack complex
R G DiScipio et al.
The Journal of biological chemistry, 263(1), 549-560 (1988-01-05)
The molecular architecture of human complement component C7 was elucidated at several structural levels. The complete primary structure of C7 was derived from the cDNA sequence of clones isolated from a human liver library. C7 is a mosaic protein that
Bernet S Kato et al.
Proteome science, 9, 73-73 (2011-11-19)
The advent of affinity-based proteomics technologies for global protein profiling provides the prospect of finding new molecular biomarkers for common, multifactorial disorders. The molecular phenotypes obtained from studies on such platforms are driven by multiple sources, including genetic, environmental, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service