Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3501207

Sigma-Aldrich

Anti-West Nile Virus Matrix antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

West Nile virus

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

West Nile Virus Matrix Antibody: West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins. However, when the viruses are inside of infected cells, the matrix protein exists in its "pre-M" form as a heterodimer with the envelope proteins. Cleavage of the "pre-M" protein to its mature form occurs during release of the virus; this cleavage leas to the dissociation of the heterodimers. The WNV receptor has recently been identified as alpha v beta 3 integrin.

Immunogen

West Nile virus matrix antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of the West Nile virus matrix protein.The immunogen is located within amino acids 220 - 270 of West Nile Virus Matrix.

Application

West Nile virus matrix antibody can be used for the detection of the West Nile virus matrix protein in ELISA. It will detect 10 ng of free peptide at 1 μg/mL.  

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service