Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB3500213

Sigma-Aldrich

Anti-EndoG antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 35 kDa

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ENDOG(2021)

General description

ENDOG (Endonuclease G) is a 28kDa mitochondria-localized protein mapped to human chromosome 9q34.1. However, in presence of certain conditions it can translocate to the nucleus through the BNIP3 (Bcl-2/adenovirus E1B 19kDa protein-interacting protein 3) cell death pathway.

Immunogen

EndoG antibody was raised with a synthetic peptide corresponding to 15 amino acids near the amino terminus of human EndoG.

Application

Anti-EndoG antibody produced in rabbit is suitable for immunohistochemistry, indirect ELISA and western blot analysis.

Biochem/physiol Actions

ENDOG (Endonuclease G) is a mitochondrial apoptotic DNase involved in the BNIP3 (Bcl-2/adenovirus E1B 19kDa protein-interacting protein 3) cell death pathway. During mitochondrial release and nuclear translocation of EndoG, BNIP3 interacts with the voltage-dependent anion channel (VDAC) to facilitate the process in the nucleus, it cleaves chromatin apoptotic DNA without the need of caspase activity. ENDOG causes DNA fragmentation during and after apoptosis. It is linked with cardiac hypertrophy and Parkinson′s disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500213.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dae Song Jang et al.
DNA and cell biology, 34(2), 92-100 (2014-11-18)
Apoptotic endonuclease G (EndoG) is responsible for DNA fragmentation both during and after cell death. Previous studies demonstrated that genetic inactivation of EndoG is cytoprotective against various pro-apoptotic stimuli; however, specific inhibitors for EndoG are not available. In this study
Xiaosha Zhang et al.
PloS one, 9(12), e113642-e113642 (2014-12-02)
BNIP3 is a proapoptotic protein that induces cell death through a mitochondria-mediated pathway. We reported previously that mitochondrial localization of BNIP3 and translocation of EndoG from mitochondria to the nucleus are critical steps of the BNIP3 pathway. It is not
V Tiranti et al.
Genomics, 25(2), 559-564 (1995-01-20)
By using a PCR-based screening of a somatic cell hybrid panel and FISH, we have assigned the loci of mitochondrial single-stranded DNA-binding protein (SSBP), mitochondrial transcription factor A (TCF6), and mitochondrial endonuclease G (ENDOG) genes to human chromosomes 7q34, 10q21
Tung Chao et al.
Autophagy, 17(11), 3444-3460 (2021-01-20)
Genotoxic insult causes nuclear and mitochondrial DNA damages with macroautophagy/autophagy induction. The role of mitochondrial DNA (mtDNA) damage in the requirement of autophagy for nuclear DNA (nDNA) stability is unclear. Using site-specific DNA damage approaches, we show that specific nDNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service