Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2702381

Sigma-Aldrich

Monoclonal Anti-C/EBP beta antibody produced in mouse

clone 21A1, affinity isolated antibody

Synonym(s):

C/EBP-beta, IL6DBP, NF-IL6, TCF5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

21A1, monoclonal

form

buffered aqueous solution

mol wt

45-48.kDa

species reactivity

human

concentration

1mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEBPB(1051)

Immunogen

The immunogen used to generate this antibody corresponds to C/EBP beta

Application

Optimal dilutions/concentrations should be determined by the researcher.

Biochem/physiol Actions

NF-IL6 is a member of the CAAT/enhancer-binding protein (C/EBP) family of transcription factors. It has been shown to directly interact with the retinoblastoma (RB) protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Protein G Affinity Purified

Physical form

Phosphate-buffered saline, pH 7.4, containing no preservatives

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Artchaya Hassametto et al.
Asian Pacific journal of cancer prevention : APJCP, 24(11), 3825-3835 (2023-11-29)
To investigate oxidative stress-related CAF transformation through C/EBPβ, which affects CRC progression and may have a potential implication for CRC treatment. The conditioned media (CM) from HCT116, CRC cells, was used to activate CCD-18Co, colon fibroblasts, then the ability of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service