Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1098

Sigma-Aldrich

Anti-Brn3a Antibody, clone 4L5, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-Anti-ATITHS, Anti-Anti-BRN3A, Anti-Anti-Oct-T1, Anti-Anti-RDC-1, Anti-Anti-brn-3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4L5, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 42.70 kDa
observed mol wt ~48 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable (peptide)

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... POU4F1(5457)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 4L5 ZooMAb is a rabbit recombinant monoclonal antibody that specifically detects Brain-specific homeobox/POU domain protein 3A, (Brn-3a).

Immunogen

KLH-conjugted linear peptide corresponding to 19 amino acids from the internal region of human homeobox/POU domain protein 3A (Brn-3a).

Application

Anti-Brn3a, clone 4L5 ZooMAb , Cat. No. ZRB1098, is a highly specific rabbit recombinant monoclonal antibody antibody that targets Brn-3a and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: 1:1,000-1:10,000 from a representative lot detected Brn3a in mouse and rat brain tissue lysates

Immunocytochemistry Analysis: A 1:40 dilution from a representative lot detected Brn3a in Neuro2A cell line.

Immunohistochemistry (Paraffin) Analysis: A 1:10,000 dilution from a representative lot detected Brn3a in human and mouse retina tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

POU domain, class 4, transcription factor 1 (UniProt: Q01851; also known as Brain-specific homeobox/POU domain protein 3A, Brain-3A, Brn-3A, Homeobox/POU domain protein RDC-1, Oct-T1) is encoded by the POU4F1 (also known as BRN3A, RDC1) gene (Gene ID: 5457) in human. Brn-3A is a multifunctional transcription factor with different regions mediating its different effects. It regulates the expression of specific genes involved in differentiation and survival within a subset of neuronal lineages. It is expressed in the brain and the retina and is present in the developing brain, spinal cord and eye. Its expression peaks early in embryogenesis and is undetectable 14 days after birth. Its C-terminal domain can act as both DNA-binding domain and a transcriptional activator. The N-terminal domain is also required for transactivation activity on some target genes acting as a discrete activation domain. Brn-3A acts by binding to sequences related to the consensus octamer motif 5′-ATGCAAAT-3′ in the regulatory regions of its target genes. The POU-specific domain of Brn-3A is localized to amino acids 261-338. Brn-3A is shown to activate Bcl-2 expression and protect neuronal cells from apoptosis. Two isoforms of Brn-3A have been described that are produced by alternative splicing. Isoform 2 can act as a transcription factor but cannot regulate the expression of the same subset of genes and does not have any anti-apoptotic effect on neuronal cells. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service