Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2701683

Sigma-Aldrich

Anti-th antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

th, tyrosine hydroxylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

zebrafish

technique(s)

western blot: 1:500-1:3000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

zebrafish ... th(30384)

General description

Tyrosine hydroxylase (TH) is a cytoplasmic enzyme. The TH gene codes for a monooxygenase.

Immunogen

Recombinant protein fragment contain a sequence corresponding to a region within amino acids 111 and 450 of th according to NP_571224

Application

Anti-th antibody produced in rabbit has been used in:
  • western blotting (1:1000)
  • whole retinal flat-mount immunolabelling (1:500)
  • immunofluorescence (2:1000)
  • immunohistochemistry (2:1000)

Biochem/physiol Actions

Cell surface-associated protein is implicated in virulence. Promotes bacterial attachment exclusively to the γ-chain of human fibrinogen. Induces formation of bacterial clumps, which diminish the ability of group IIA phospholipase A2 to cause bacterial phospholipid hydrolysis and killing. Significantly decreases macrophage phagocytosis possibly thanks to the clumps, clumped bacteria being too large to be phagocytosed. Dominant factor responsible for human platelet aggregation, which may be an important mechanism for initiating infective endocarditis. Enhances spleen cell proliferative response in vitro, contributing significantly to the immunostimulatory activity of S.aureus.
Tyrosine hydroxylase (TH), the rate-limiting enzyme of dopamine synthesis, is essential for catecholamine biosynthesis. Its major role is to catalyze the conversion of L-tyrosine to L-dihydroxyphenylalanine (L-DOPA). TH mainly serves as marker furcate cholaminergic neurons. Missense mutations in the TH gene are associated with Segawa′s syndrome, L-DOPA responsive infantile Parkinsonism, or L-DOPA responsive dystonia (DRD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 1% BSA, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soundhar Ramasamy et al.
The Journal of experimental biology, 222(Pt 1) (2018-11-18)
High fecundity, transparent embryos for monitoring the rapid development of organs and the availability of a well-annotated genome has made zebrafish a model organism of choice for developmental biology and neurobiology. This vertebrate model, which is also a favourite in
Chen Zhu et al.
Fitoterapia, 117, 47-51 (2017-01-09)
In this study, to study the effect of rhynchophylline on TH in midbrain of methamphetamine-induced conditioned place preference (CPP) adult zebrafish, place preference adult zebrafish models were established by methamphetamine (40μg/g) and the expression of TH was observed by immunohistochemistry
Guney Bademci et al.
CNS & neurological disorders drug targets, 11(4), 469-481 (2012-05-16)
The tyrosine hydroxylase (TH) gene encodes a monoxygenase that catalyzes the rate limiting step in dopamine biosynthesis. A hallmark of Parkinson's disease (PD) is the loss of dopaminergic neurons in the substantia nigra. Consistent with the essential role of TH
Dong-Jun Lv et al.
Behavioural brain research, 372, 112031-112031 (2019-06-14)
Parkinson's disease (PD) is the second most common neurodegenerative disorder in the world. Apart from motor deficits, PD reduces patient's quality of life through sleep disturbances, cognitive impairment and emotional disorders. However, it's unclear whether bad life habits such as
Constance Pierre et al.
The Journal of experimental biology, 223(Pt 18) (2020-08-02)
The neurotransmitter serotonin controls a variety of physiological and behavioral processes. In humans, mutations affecting monoamine oxidase (MAO), the serotonin-degrading enzyme, are highly deleterious. Yet, blind cavefish of the species Astyanax mexicanus carry a partial loss-of-function mutation in MAO (P106L)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service