Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2500826

Sigma-Aldrich

Anti-Proopiomelanocortin/POMC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ACTH, Anti-CLIP, Anti-POC

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POMC(5443)

General description

Proopiomelanocortin (POMC) is a pro-peptide and multifunctional precursor protein for melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone ACTH. The POMC gene is mapped to human chromosome 2p23.3. It is expressed in skin, pituitary gland, and tissues of the immune system.

Immunogen

Peptide with sequence C-NAIIKNAYKKGE from the C Terminus of the protein sequence according to NP_000930.1; NP_001030333.1.

Biochem/physiol Actions

Proopiomelanocortin (POMC) expression is regulated by hypermethylation of the upstream CpG-rich island in the gene. Mutations in the POMC gene are implicated in adrenocorticotropic hormone (ACTH) deficiency. Adrenal insufficiency and obesity are correlated to loss of function mutations in POMC.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Böhm et al.
Endocrine reviews, 33(4), 623-651 (2012-06-28)
Proopiomelanocortin (POMC)-derived peptides such as melanocortins and β-endorphin (β-ED) exert their pleiotropic effects via binding to melanocortin receptors (MCR) and opioid receptors (OR). There is now compelling evidence for the existence of a functional POMC system within the osteoarticular system.
Meenal S Mendiratta et al.
International journal of pediatric endocrinology, 2011(1), 5-5 (2011-08-24)
Isolated hypocortisolism due to ACTH deficiency is a rare condition that can be caused by homozygous or compound heterozygous mutations in the gene encoding proopiomelanocortin (POMC). Loss of function mutations of POMC gene typically results in adrenal insufficiency, obesity and
Rosa Gallo et al.
iScience, 27(7), 110259-110259 (2024-07-19)
Overeating leads to obesity, a low-grade inflammatory condition involving interleukin-17A (IL-17A). While pro-opiomelanocortin (POMC) neurons regulate feeding, their connection with IL-17A is not well understood. To impair IL-17A signaling in POMC neurons, IL-17A receptor (Il17ra) was deleted by crossing IL17ra-flox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service