Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2109106

Sigma-Aldrich

Anti-TOB2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

TOB4, TOBL, TROB2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity (predicted by homology)

rabbit, mouse, guinea pig, zebrafish, sheep, bovine, human, goat, canine, horse

concentration

0.5 mg/mL

technique(s)

western blot: 1 μg/mL

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOB2(57396)

General description

TOB2 belongs to the TOB/BTG1 family of antiproliferative proteins, which are involved in the regulation of cell cycle progression.

Immunogen

Synthetic peptide directed towards the middle region of Human TOB2

Sequence

Synthetic peptide located within the following region: EGHWYPEKPLKGSGFRCVHIGEMVDPVVELAAKRSGLAVEDVRANVPEEL

Physical form

Supplied at 0.5 mg/ml in phosphate-buffered saline, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service