Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
11101706
EC Number:
231-111-4
MDL number:
Quality Level
assay
≥99% trace metals basis
form
nanopowder
resistivity
6.97 μΩ-cm, 20°C
avg. part. size
<100 nm
bp
2732 °C (lit.)
mp
1453 °C (lit.)
density
8.9 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
[Ni]
InChI
1S/Ni
InChI key
PXHVJJICTQNCMI-UHFFFAOYSA-N
Application
Nickel nanopowder is a magnetic material, which has a variety of usages such as:
- catalytic applications
- hydrogen transfer reactions
- contrasting agent in biological systems
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Carc. 2 - Skin Sens. 1 - STOT RE 1 Inhalation
target_organs
Respiratory Tract,Skin
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
This thematic issue focuses on the emerging applications of nanomaterials. Nanomaterials are defined as substances with at least one dimension smaller than 100 nm.
Nickel nanoparticles obtained by a modified polyol process: synthesis, characterization, and magnetic properties
Couto GG, et al.
Journal of Colloid and Interface Science, 311(2), 461-468 (2007)
Nickel nanoparticles in hydrogen transfer reactions
Alonso F, et al.
Accounts of Chemical Research, 44(5), 379-391 (2011)
E R Leite et al.
Journal of nanoscience and nanotechnology, 2(1), 89-94 (2003-08-12)
Here we describe a new route for the synthesis of nanometric Ni particles embedded in a mesoporous silica material with excellent potential for catalytic applications. Mesoporous silica with a surface area in the range of 202-280 m2/g, with narrow pore
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 577995-50G | 04065273496925 |
| 577995-5G | 04061832629018 |

