Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1412101

Sigma-Aldrich

ANTI-TOP3B antibody produced in mouse

clone 4H7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ39376, TOP3B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4H7, monoclonal

form

buffered aqueous solution

mol wt

antigen 120.56 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOP3B(8940)

General description

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus relaxing the supercoils and altering the topology of DNA. The enzyme interacts with DNA helicase SGS1 and plays a role in DNA recombination, cellular aging and maintenance of genome stability. Alternative splicing of the C-terminus of this gene results in three transcript variants which have distinct tissue specificity; however, not all variants have been fully described. (provided by RefSeq)

Immunogen

TOP3B (AAH02432, 1 a.a. ~ 862 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MKTVLMVAEKPSLAQSIAKILSRGSLSSHKGLNGACSVHEYTGTFAGQPVRFKMTSVCGHVMTLDFLGKYNKWDKVDPAELFSQAPTEKKEANPKLNMVKFLQVEGRGCDYIVLWLDCDKEGENICFEVLDAVLPVMNKAHGGEKTVFRARFSSITDTDICNAMACLGEPDHNEALSVDARQELDLRIGCAFTRFQTKYFQGKYGDLDSSLISFGPCQTPTLGFCVERHDKIQSFKPETYWVLQAKVNTDKDRSLLLDWDRVRVFDREIAQMFLNMTKLEKEAQVEATSRKEKAKQRPLALNTVEMLRVASSSLGMGPQHAMQTAERLYTQGYISYPRTETTHYPENFDLKGSLRQQANHPYWANTVKRLLAEGINRPRKGHDAGDHPPITPMKSATEAELGGDAWRLYEYITRHFIATVSHDCKYLQSTISFRIGPELFTCSGKTVLSPGFTEVMPWQSVPLEESLPTCQRGDAFPVGEVKMLEKQTNPPDYLTEAELITLMEKHGIGTDASIPVHINNICQRNYVTVESGRRLKPTNLGIVLVHGYYKIDAELVLPTIRSAVEKQLNLIAQGKADYRQVLGHTLDVFKRKFHYFVDSIAGMDELMEVSFSPLAATGKPLSRCGKCHRFMKYIQAKPSRLHCSHCDETYTLPQNGTIKLYKELRCPLDDFELVLWSSGSRGKSYPLCPYCYNHPPFRDMKKGMGCNECTHPSCQHSLSMLGIGQCVECESGVLVLDPTSGPKWKVACNKCNVVAHCFENAHRVRVSADTCSVCEAALLDVDFNKAKSPLPGDETQHMGCVFCDPVFQELVELKHAASCHPMHRGGPGRRQGRGRGRARRPPGKPNPRRPKDKMSALAAYFV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service