Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1401757

Sigma-Aldrich

Anti-FEM1A antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp762M136, EPRAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FEM1A(55527)

Immunogen

FEM1A (NP_061178.1, 1 a.a. ~ 669 a.a) full-length human protein.

Sequence
MDLRTAVYNAARDGKLQLLQKLLSGRSREELDELTGEVAGGGTPLLIAARYGHLDVVEYLVDRCGASVEAGGSVHFDGETIEGAPPLWAASAAGHLDVVRSLLRRGASVNRTTRTNSTPLRAACFDGHLEVVRYLVGEHQADLEVANRHGHTCLMISCYKGHREIARYLLEQGAQVNRRSAKGNTALHDCAESGSLEILQLLLGCKARMERDGYGMTPLLAASVTGHTNIVEYLIQEQPGQEQVAGGEAQPGLPQEDPSTSQGCAQPQGAPCCSSSPEEPLNGESYESCCPTSREAAVEALELLGATYVDKKRDLLGALKHWRRAMELRHQGGEYLPKPEPPQLVLAYDYSREVNTTEELEALITDPDEMRMQALLIRERILGPSHPDTSYYIRYRGAVYADSGNFERCIRLWKYALDMQQSNLEPLSPMTASSFLSFAELFSYVLQDRAAKGSLGTQIGFADLMGVLTKGVREVERALQLPREPGDSAQFTKALAIILHLLYLLEKVECTPSQEHLKHQTVYRLLKCAPRGKNGFTPLHMAVDKDTTNVGRYPVGRFPSLHVVKVLLDCGADPDSRDFDNNTPLHIAAQNNCPAIMNALIEAGAHMDATNAFKKTAYELLDEKLLARGTMQPFNYVTLQCLAARALDKNKIPYKGFIPEDLEAFIELH

Biochem/physiol Actions

FEM1A (fem-1 homolog A) protein is involved in the regulation of stem-loop binding protein, that is associated with mRNA cleavage, translation and degradation. FEM1A is involved in anti-inflammatory signaling by interacting with prostaglandin E2 subtype E4 receptors. FEM1A is associated with the pathogenesis of polycystic ovary syndrome. Studies show that FEM1A gene is downegulated in rhabdomyosarcoma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Fem1a gene is downregulated in Rhabdomyosarcoma.
Ventura-Holman T
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 26(6), 294-299 (2005)
FEM1 proteins are ancient regulators of SLBP degradation.
Dankert
Cell Cycle, 16(6), 556-564 (2017)
M O Goodarzi et al.
Human reproduction (Oxford, England), 23(12), 2842-2849 (2008-09-02)
Human homologs (FEM1A, FEM1B, FEM1C) of nematode sex determination genes are candidate genes for polycystic ovary syndrome (PCOS). We previously identified a FEM1A mutation (H500Y) in a woman with PCOS; FEM1B has been implicated in insulin secretion. Women with and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service