Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1301597

Sigma-Aldrich

ANTI-AVP (CENTER) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

ARVP, AVP, VP, Vasopressin-neurophysin 2-copeptin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

17325 Da

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AVP(551)

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liem T Bui et al.
Journal of experimental botany, 70(6), 1815-1827 (2019-03-13)
Ethanol fermentation is considered as one of the main metabolic adaptations to ensure energy production in higher plants under anaerobic conditions. Following this pathway, pyruvate is decarboxylated and reduced to ethanol with the concomitant oxidation of NADH to NAD+. Despite
Meng-Syuan Lin et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(8), 1505-1524 (2018-12-28)
Intrinsically photosensitive retinal ganglion cells (ipRGCs), which express the photopigment melanopsin, are photosensitive neurons in the retina and are essential for non-image-forming functions, circadian photoentrainment, and pupillary light reflexes. Five subtypes of ipRGCs (M1-M5) have been identified in mice. Although
Emma Kneuss et al.
Genes & development, 33(17-18), 1208-1220 (2019-08-17)
The PIWI-interacting RNA (piRNA) pathway is a conserved small RNA-based immune system that protects animal germ cell genomes from the harmful effects of transposon mobilization. In Drosophila ovaries, most piRNAs originate from dual-strand clusters, which generate piRNAs from both genomic
Chao-Hung Chen et al.
Biochimica et biophysica acta. General subjects, 1863(6), 1059-1069 (2019-03-25)
O-GlcNAcylation is an important mechanism of diabetic complication. Retinoid homeostasis regulates cell-physiological functions through STRA6-retinol signaling. Therefore, we investigated whether O-GlcNAcylation disrupted STRA6-retinol signals in diabetes. Immunoprecipitation and proximity ligation assay were used to investigate O-GlcNAcylation of STRA6-retinol signals in
Tom Flossmann et al.
Cell reports, 26(12), 3173-3182 (2019-03-21)
Synchronized activity is a universal characteristic of immature neural circuits that is essential for their developmental refinement and strongly depends on GABAergic neurotransmission. A major subpopulation of GABA-releasing interneurons (INs) expresses somatostatin (SOM) and proved critical for rhythm generation in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service