Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV42196

Sigma-Aldrich

Anti-ITGBL1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Integrin, β-like 1 (with EGF-like repeat domains), Anti-OSCP, Anti-TIED

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

horse, pig, dog, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGBL1(9358)

Related Categories

General description

Integrin, β-like 1 (with EGF-like repeat domains) (ITGBL1, TIED) is a β integrin-related protein found in aorta, thymus and osteogenic sarcoma. Defects in ITGBL1 may be associated with isolated growth hormone deficiency.

Specificity

Anti-ITGBL1 (AB1) polyclonal antibody reacts with canine, human, bovine, mouse, and rat integrin, β-like 1 (with EGF-like repeat domains) proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human ITGBL1

Application

Anti-ITGBL1 (AB1) polyclonal antibody is used to tag integrin, β-like 1 (with EGF-like repeat domains) protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of integrin, β-like 1 (with EGF-like repeat domains) in cell processes such as isolated growth hormone deficiency

Sequence

Synthetic peptide located within the following region: MRPPGFRNFLLLASSLLFAGLSAVPQSFSPSLRSWPGAACRLSRAESERR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service