Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300971

Sigma-Aldrich

Anti-PIK3R2 (Y467) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PI3-kinase p85 subunit β, Anti-PIK3R2, Anti-Phosphatidylinositol 3-kinase regulatory subunit β, Anti-PtdIns-3-kinase p85-β

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr467)

Gene Information

human ... PIK3R2(5296)

General description

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. PIK3R2 binds to activated Protein Tyrosine Kinases, which are phosphorylated, through its SH2 domain, and acts as an adaptor, mediating the association of the P110 catalytic unit to the plasma membrane.

Immunogen

PIK3R2 (NP_005018, )
This antibody is generated from rabbits immunized with a peptide corresponding to amino acid residues surrounding Y467 of human PIK3R2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service