Skip to Content

Legal compliance reminder: purchasers must obtain all necessary licenses and comply with applicable laws (see General Terms & Conditions of Sale). We reserve the right to request additional information.

MilliporeSigma

Skip To

S8777

Worthington Accessories

tipping stand for 25 L Dewar

Synonym(s):

dewar stand, D025-8C00, cryogenic dewar, cryogenic storage, dewar tipping stand

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.43
UNSPSC Code:
41100000
Packaging:
pack of 1 ea
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pack of 1 ea

manufacturer/tradename

Worthington 366582

technique(s)

cryopreservation: suitable

General description

This stand supports the Dewar and allows convenient, safe pouring of liquid. Wheels add mobility and additional handling ease.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R0384Z744000Z744003
description

tipping stand for 25 L Dewar

description

5 shelf rack for LS750 Dewar

description

rack for 10 drawers for LS6000 liquid nitrogen refrigerator

description

necktube core for LS3000 liquid nitrogen refrigerator

manufacturer/tradename

Worthington 366582

manufacturer/tradename

Worthington R036-9C34

manufacturer/tradename

Worthington RS60-9C50

manufacturer/tradename

Worthington RS30-9C16

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

technique(s)

cryopreservation: suitable

technique(s)

cryopreservation: suitable

technique(s)

cryopreservation: suitable

technique(s)

cryopreservation: suitable


Still not finding the right product?

Explore all of our products under Worthington Accessories




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library