Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

S4019

Sigma-Aldrich

Sodium bicarbonate

Hybri-Max, powder, suitable for hybridoma, ≥99.5%

Synonym(s):

Sodium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein:
4153970
EC Number:
MDL number:
UNSPSC Code:
12161702
PubChem Substance ID:
NACRES:
NA.75

grade

Hybri-Max

Assay

≥99.5%

form

powder

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

pH

8.3 (0.1 molfreshly prepared)

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

solubility

water: soluble 100 mg/mL

density

2.16 g/mL at 25 °C (lit.)

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium bicarbonate has been used as a supplement:
  • in minimum essential medium (MEM)/flushing medium for embryo isolation
  • in TCM 199 medium for the collection of cumulus-oocyte complexes (COCs) and in vitro maturation (IVM)
  • in IVM medium for in vitro maturation of COCs

Biochem/physiol Actions

Sodium bicarbonate occurs naturally as the mineral nahcolite. It is used to prepare many sodium salts as well as a carbon dioxide source. Sodium bicarbonate is used in the manufacture of fire extinguishers and cleaning agents. Bicarbonate plays an important role in pH regulation in vivo. In biochemistry, molecular biology, and chromatography, sodium bicarbonate is used as a buffer. This Hybri-Max product is suitable for use in hybridoma and cell culture research (2.2 mg/ml).

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of estrous ewe serum and heparin on in vitro fertilization and subsequent embryonic development in sheep
Li F, et al.
Small Ruminant Research, 63(3), 226-232 (2006)
The effects of L-ergothioneine and L-ascorbic acid on the in vitro maturation (IVM) and embryonic development (IVC) of sheep oocytes
Development, 1, 5-14 (2010)
Shay P McGuinness et al.
Critical care medicine, 41(7), 1599-1607 (2013-05-21)
Cardiac surgery-associated acute kidney injury occurs in up to 50% of patients and is associated with increased mortality and morbidity. This study aimed to discover if perioperative urinary alkalinization with sodium bicarbonate infusion reduces the prevalence of cardiac surgery-associated acute
Joshua C Tremblay et al.
The Journal of physiology, 593(3), 723-737 (2014-11-25)
Blood flow through intrapulmonary arteriovenous anastomoses (IPAVA) is increased by acute hypoxia during rest by unknown mechanisms. Oral administration of acetazolamide blunts the pulmonary vascular pressure response to acute hypoxia, thus permitting the observation of IPAVA blood flow with minimal
Sophia Zoungas et al.
Annals of internal medicine, 151(9), 631-638 (2009-11-04)
Intravenous sodium bicarbonate has been proposed to reduce the risk for contrast-induced nephropathy (CIN). To determine the effect of sodium bicarbonate on the risk for CIN. MEDLINE, PubMed, EMBASE, and the Cochrane Central Register of Controlled Trials from 1950 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service