Skip to Content
MilliporeSigma

Skip To

S1899

N-Succinyl-Gly-Gly-Phe-p-nitroanilide

protease substrate, ≥98% (TLC), powder

Synonym(s):

Suc-Gly-Gly-Phe-PNA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C23H25N5O8
CAS Number:
Molecular Weight:
499.47
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
273-486-7
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

N-Succinyl-Gly-Gly-Phe-p-nitroanilide, protease substrate

Quality Segment

assay

≥98% (TLC)

form

powder

solubility

acetic acid: 20 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OC(=O)CCC(=O)NCC(=O)NCC(=O)NC(Cc1ccccc1)C(=O)Nc2ccc(cc2)N(=O)=O

InChI

1S/C23H25N5O8/c29-19(10-11-22(32)33)24-13-20(30)25-14-21(31)27-18(12-15-4-2-1-3-5-15)23(34)26-16-6-8-17(9-7-16)28(35)36/h1-9,18H,10-14H2,(H,24,29)(H,25,30)(H,26,34)(H,27,31)(H,32,33)

InChI key

CFYIUBWVKZQDOG-UHFFFAOYSA-N

General description

Substrate for chymotrypsin and for S. griseus protease B.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S2628S4760573462
description

protease substrate

description

protease substrate

description

elastase substrate

description

Sensitive chromogenic substrate for human leukocyte cathepsin G that is not reactive with elastase.

assay

≥98% (TLC)

assay

≥98% (TLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

solubility

acetic acid: 20 mg/mL, clear, colorless to faintly yellow

solubility

dioxane: 25 mg/mL, clear, colorless to faintly yellow

solubility

DMF: 25 mg/mL, clear, colorless to light yellow

solubility

DMSO: 100 mg/mL

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

powder

form

powder

form

powder

form

solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters