Skip to Content
MilliporeSigma
All Photos(2)

Documents

RES1513C-A7

SAFC

L-Cystine dihydrochloride, non-animal derived

Pharma Manufacturing

Synonym(s):

L-Cystine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O4S2 · 2HCl
CAS Number:
Molecular Weight:
313.22
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.21

biological source

synthetic

Quality Level

description

Non-Animal Derived

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, microbial, and trace metals, tested

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

Cl.Cl.N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2.2ClH/c7-3(5(9)10)1-13-14-2-4(8)6(11)12;;/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12);2*1H/t3-,4-;;/m0../s1

InChI key

HHGZUQPEIHGQST-RGVONZFCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Application

L-Cystine is a sulfur containing non-essential amino acid. It is used as a cell culture media component and raw material in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-Cystine dihydrochloride is a modified version of L-Cystine with improved solubility.

Packaging

This product is available in the following package sizes:
RES1513C-A701X: 100g container
RES1513C-A704X: 10kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service