Skip to Content
MilliporeSigma
All Photos(1)

Documents

137253

SAFC

L-Cystine dihydrochloride

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

L-Cystine Dihydrochloride, Non-Animal Derivative

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O4S2 · 2HCl
CAS Number:
Molecular Weight:
313.22
MDL number:
UNSPSC Code:
12352209

biological source

synthetic

Quality Level

description

Non-Animal Derived

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and trace metals, tested

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

storage temp.

2-30°C

SMILES string

Cl.Cl.N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2.2ClH/c7-3(5(9)10)1-13-14-2-4(8)6(11)12;;/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12);2*1H/t3-,4-;;/m0../s1

InChI key

HHGZUQPEIHGQST-RGVONZFCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Cystine is a sulfur containing non-essential amino acid. It is used as a cell culture media component and raw material in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-Cystine dihydrochloride is a modified version of L-Cystine with improved solubility.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service