Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RES1164B-A7

SAFC

BIS-TRIS hydrochloride

Pharma Manufacturing

Synonym(s):

BIS-TRIS hydrochloride, 2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)aminotris(hydroxymethyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19NO5 · HCl
CAS Number:
Molecular Weight:
245.70
MDL number:
UNSPSC Code:
12161705
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin and microbial; tested

useful pH range

5.8-7.2

pKa 

6.5

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

Cl.OCCN(CCO)C(CO)(CO)CO

InChI

1S/C8H19NO5.ClH/c10-3-1-9(2-4-11)8(5-12,6-13)7-14;/h10-14H,1-7H2;1H

InChI key

VEYRVLHAMHQVTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

Bis Tris HCl is a commonly used biological buffer for very broad usage in biological systems such as diagnostics, electrophoresis and protein based bioprocessing. The pKa of Bis Tris is 6.36, which makes it a good candidate for biological-based formulations.

Bis Tris HCl can be used in a wide variety of biological applications including culture media, electrophoresis separations and diagnostic reagent formulations.

Packaging

Product is available in the following package sizes:
RES1164B-A701X: 100 gm container
RES1164B-A702X: 1 kg container
RES1164B-A704X: 10 kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service