Skip to Content
MilliporeSigma

L5777

N-Lauroylsarcosine sodium salt

BioXtra, ≥97% (TLC)

Synonym(s):

N-Dodecanoyl-N-methylglycine sodium salt, Sarkosyl NL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
205-281-5
Beilstein/REAXYS Number:
5322974
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

anionic

Quality Segment

product line

BioXtra

assay

≥97% (TLC)

form

powder

mol wt

micellar avg mol wt 600, average mol wt 600

aggregation number

2

impurities

≤0.1% Insoluble matter, ≤0.1% Phosphorus (P)

CMC

14.6 mM (20-25°C)

solubility

H2O: 0.1 M, clear, colorless to very faintly yellow

density

(1,14 g/cm3 at 20 °C )

cation traces

Al: ≤0.0005%, Ca: ≤0.01%, Cu: ≤0.0005%, Fe: ≤0.005%, K: ≤0.01%, Mg: ≤0.005%, NH4+: ≤0.05%, Pb: ≤0.01%, Zn: ≤0.0005%

SMILES string

[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

General description

N-Lauroylsarcosine sodium salt is an anionic surfactant.

Application

N-Lauroylsarcosine sodium salt has been used in a study to assess involvement of histamine released from epidermal keratinocytes. It has also been used in a study to develop a simple method for extracting DNA from Cryptosporidium oocysts.

Other Notes

Anionic surfactant.


Still not finding the right product?

Explore all of our products under N-Lauroylsarcosine sodium salt


pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 1

flash_point_f

512.6 °F - closed cup

flash_point_c

267 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library