Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB1162

Sigma-Aldrich

Rat Renin-1 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

rat

technique(s)

ELISA: suitable

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 68 pg/ml

shipped in

wet ice

storage temp.

−20°C

General description

This ELISA antibody pair detects Rat Renin (REN)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marite Punapart et al.
Genes, 12(11) (2021-11-28)
Wolfram syndrome (WS) 1 is a rare monogenic neurodegenerative disorder caused by mutations in the gene encoding WFS1. Knowledge of the pathophysiology of WS is incomplete and to date, there is no treatment available. Here, we describe early deviations in
J Oh et al.
Nature communications, 11(1), 4798-4798 (2020-09-25)
Myeloid cells are known mediators of hypertension, but their role in initiating renin-induced hypertension has not been studied. Vitamin D deficiency causes pro-inflammatory macrophage infiltration in metabolic tissues and is linked to renin-mediated hypertension. We tested the hypothesis that impaired
Junda Hu et al.
Acta physiologica (Oxford, England), 232(1), e13612-e13612 (2020-12-31)
The use of calcineurin inhibitors such as cyclosporine A (CsA) for immunosuppression after solid organ transplantation is commonly limited by renal side effects. CsA-induced deterioration of glomerular filtration rate and sodium retention may be related to juxtaglomerular dysregulation as a
Dong Cheng et al.
Molecular medicine reports, 22(5), 4151-4162 (2020-10-02)
Clinical application of doxorubicin (DOX) is hampered by its potential cardiotoxicity, however angiotensin receptor blockers could attenuate DOX‑induced cardiomyopathy. The present study tested the hypothesis that simultaneous administration of valsartan (Val) with DOX could prevent DOX‑induced myocardial injury by modulating myocardial NAD(P)H

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service