Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB1074

Sigma-Aldrich

Human PGRN ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum sample(s)
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.1 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GRN(2896)

General description

This ELISA antibody pair detects human PGRN. Other species are not determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anfei Huang et al.
JCI insight, 4(17) (2019-09-06)
`NK cell-mediated regulation of antigen-specific T cells can contribute to and exacerbate chronic viral infection, but the protective mechanisms against NK cell-mediated attack on T cell immunity are poorly understood. Here, we show that progranulin (PGRN) can reduce NK cell
Yanqing Li et al.
Journal of neuropathology and experimental neurology, 78(5), 406-415 (2019-04-03)
Progranulin (PGRN) plays critical roles in inflammation, tumorigenesis, and neurodegeneration. PGRN levels in blood and cerebrospinal fluid (CSF) are being increasingly investigated as potential biomarkers for these disorders. However, the value of CSF PGRN as a biomarker has been limited
Yingguang Han et al.
Inflammation, 40(3), 946-955 (2017-03-02)
The aim of this study was to investigate progranulin (PGRN) expression and its effect in cartilage degradation and in the pathogenesis of osteonecrosis of the femoral head (ONFH). Cartilage specimens were obtained from ONFH and FNF patients and PGRN expression
Wei Xu et al.
Science advances, 6(43) (2020-10-23)
Progranulin (PGRN) is a secreted pleiotropic glycoprotein associated with the development of common neurodegenerative diseases. Understanding the pathophysiological role of PGRN may help uncover biological underpinnings. We performed a genome-wide association study to determine the genetic regulators of cerebrospinal fluid
Lianlian Liu et al.
BMC immunology, 21(1), 32-32 (2020-06-07)
Macrophage M1 polarization plays a pivotal role in inflammatory diseases. Progranulin (PGRN) has potential anti-inflammation action, however, the effect of PGRN on macrophage M1 polarization has been poorly studied. Our study aimed to investigate the effect of PGRN on lipopolysaccharide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service