MilliporeSigma
  • The FAM171A2 gene is a key regulator of progranulin expression and modifies the risk of multiple neurodegenerative diseases.
The FAM171A2 gene is a key regulator of progranulin expression and modifies the risk of multiple neurodegenerative diseases.
Science advances (2020-10-23)
Wei Xu, Si-Da Han, Can Zhang, Jie-Qiong Li, Yan-Jiang Wang, Chen-Chen Tan, Hong-Qi Li, Qiang Dong, Cui Mei, Lan Tan, Jin-Tai Yu
ABSTRACT
Progranulin (PGRN) is a secreted pleiotropic glycoprotein associated with the development of common neurodegenerative diseases. Understanding the pathophysiological role of PGRN may help uncover biological underpinnings. We performed a genome-wide association study to determine the genetic regulators of cerebrospinal fluid (CSF) PGRN levels. Common variants in region of FAM171A2 were associated with lower CSF PGRN levels (rs708384, P = 3.95 × 10-12). This was replicated in another independent cohort. The rs708384 was associated with increased risk of Alzheimer's disease, Parkinson's disease, and frontotemporal dementia and could modify the expression of the FAM171A2 gene. FAM171A2 was considerably expressed in the vascular endothelium and microglia, which are rich in PGRN. The in vitro study further confirmed that the rs708384 mutation up-regulated the expression of FAM171A2, which caused a decrease in the PGRN level. Collectively, genetic, molecular, and bioinformatic findings suggested that FAM171A2 is a key player in regulating PGRN production.
Sigma-Aldrich
Human PGRN ELISA Kit, for cell culture supernatants, plasma, and serum samples