Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0617

Sigma-Aldrich

Human APOH / Beta-2-glycoprotein 1 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 30 pg/mL
standard curve range: 24.58-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... APOH(350)

General description

The ApoH ELISA kit provides for the Quantitative measurement of ApoH in Cell Culture Supernatants, Plasma and Serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel Serrano et al.
Frontiers in immunology, 10, 1031-1031 (2019-05-28)
Background: Antiphospholipid syndrome (APS) is characterized by thrombosis and/or pregnancy morbidity with presence of anti-phospholipid antibodies (aPL). The APS classification criteria only consider the aPL of IgG/IgM isotype, however testing of aPL of IgA isotype is recommended when APS is
Gavriella-Zoi Vrakopoulou et al.
In vivo (Athens, Greece), 34(5), 2423-2429 (2020-09-03)
Healing is related to gastrointestinal anastomotic leak, which is a severe and common complication. This study aimed to investigate the feasibility and the impact of deserosalization on healing of jejuno-jejunal anastomoses in an animal model. Seven swine underwent three types
Shuo Chen et al.
Chemosphere, 226, 192-200 (2019-03-31)
Exogenous ammonium nitrogen (AN) fertilization/deposition and the subsequently induced soil acidification, might exacerbate soil phosphorus (P) transformation and mobility, whereas poor understanding in abiotic and biotic processes still existed between intensively cultivated soil with high proportion of legacy inorganic P
Pradeep K Kudipudi et al.
International journal of molecular sciences, 20(24) (2019-12-15)
Transforming growth factor-βs (TGF-βs) signal after binding to the TGF-β receptors TβRI and TβRII. Recently, however, betaglycan (BG) was identified as an important co-receptor, especially for TGF-β2. Both proteins are involved in several testicular functions. Thus, we analyzed the importance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service