Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0287

Sigma-Aldrich

Mouse IL-2 ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

IL-1 beta, Interleukin-1 beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 0.625-40 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il2(16183)

General description

The Mouse IL-2 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IL-2 in serum, plasma and cell culture supernatants.

Immunogen

Recombinant Mouse IL-2

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shanji Nan et al.
Molecular therapy. Nucleic acids, 24, 646-657 (2021-05-14)
MicroRNA (miR)-410 plays a potential role in the pathogenesis of atherosclerosis. The current study mainly focuses on the underlying mechanism of miR-410/histone deacetylase 1 (HDAC1)/KLF5/nuclear factor κB (NF-κB) inhibitor α (IKBα)/NF-κB axis in atherosclerosis. miR-410 expression was determined using quantitative real-time
Ying Wang et al.
Journal of cellular and molecular medicine, 24(11), 6107-6119 (2020-04-24)
Non-alcoholic fatty liver disease (NAFLD) serves as the most common subtype of liver diseases and cause of liver dysfunction, which is closely related to obesity and insulin resistance. In our study, we sought to investigate effect of transcription factor grainyhead-like
Yujo Kawashita et al.
Radiation research, 182(2), 191-200 (2014-06-28)
Hepatocellular carcinoma (HCC) often presents as a diffuse or multifocal tumor making it difficult to control by surgery or radiation. Radio-inducible herpes simplex virus thymidine kinase (HSV-TK) gene therapy has been shown to enhance local tumor control after radiation therapy
Song Gao et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 63(6), 441-450 (2014-02-06)
CD200 is expressed on various cell types, including T cells, while the CD200 receptor (CD200R) is expressed on myeloid cells such as monocytes-derived macrophages (MDMs). The CD200-CD200R interaction has been shown to play an important role in the prevention of
Azita Haddadi et al.
Nanotechnology, 25(35), 355101-355101 (2014-08-15)
Mannan (MN) is the natural ligand for mannose receptors, which are widely expressed on dendritic cells (DCs). The purpose of this study was to assess the effect of formulation parameters on the immunogenicity of MN-decorated poly (D, L-lactide-co-glycolide) (PLGA) nanoparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service