Skip to Content
MilliporeSigma
All Photos(2)

Documents

RAB0263

Sigma-Aldrich

Mouse IL-17 (IL-17A) ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 6.1-600 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il17a(16171)

General description

The Mouse IL-17 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IL-17 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse IL-17

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lan Yao et al.
Immunology letters, 161(1), 76-80 (2014-05-20)
Recent data have shown that piperlongumine (PL), an important component of Piper longum fruits, is known to possess anti-inflammatory and vascular-protective activities. This study aimed to examine the therapeutic effects and underlying mechanisms of PL on lupus-prone MRL-Fas(lpr) mice. Female
Jinyu Zhang et al.
International immunology, 26(7), 407-415 (2014-03-22)
MiR-155 (-/-) mice are highly resistant to experimental autoimmune encephalomyelitis (EAE), while Pdcd1 (-/-) mice develop a more severe form of the disease. To determine the conflicting roles of these two molecules in the disease, we generated miR-155 (-/-) Pdcd1
C Keijzer et al.
Vaccine, 32(24), 2904-2910 (2014-03-07)
Nasal vaccination is considered to be a promising alternative for parenteral vaccination against influenza virus as it is non-invasive and offers the opportunity to elicit strong antigen-specific responses both systemic and locally at the port of entry of the pathogen.
Melanie Stumpf et al.
European journal of immunology, 44(6), 1737-1746 (2014-03-22)
Cytotoxic T lymphocyte antigen-4 (CTLA-4) is a major negative regulatory molecule for T-cell activation with a complex biology and function. CTLA-4 is known to regulate homeostatic lymphoproliferation as well as tolerance induction and has been proposed to be an important
Anuradha Bandaru et al.
European journal of immunology, 44(7), 2013-2024 (2014-03-20)
We studied the factors that regulate IL-23 receptor expression and IL-17 production in human tuberculosis infection. Mycobacterium tuberculosis (M. tb)-stimulated CD4(+) T cells from tuberculosis patients secreted less IL-17 than did CD4(+) T cells from healthy tuberculin reactors (PPD(+) ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service