Skip to Content
MilliporeSigma
All Photos(2)

Documents

RAB0262

Sigma-Aldrich

Human IL-17 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

Cytokine detection kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL17A(3605)

General description

The Human IL-17 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-17 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IL-17

Application

Human IL-17 ELISA Kit has been used in serum IL17 measurement.

Biochem/physiol Actions

IL-17A (interleukin 17A) plays a major role in the pathogenesis of autoimmune diseases like multiple sclerosis and rheumatoid arthritis. It also plays a pathogenic role in allergic rhinitis (AR). It helps in the formation of several chemokines and cytokines in airway epithelial cells.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum interleukin (IL)-17 in psoriasis
Mawla M A, et al.
Journal of American Science, 9(12), 229-232 (2013)
IL-17A and Th17 cells in lung inflammation: an update on the role of Th17 cell differentiation and IL-17R signaling in host defense against infection
Tsai H C, et al.
Clinical & Developmental Immunology, 2013, 647-653 (2013)
Anyamanee Chaiprasongsuk et al.
International journal of molecular sciences, 21(24) (2020-12-16)
Lumisterol (L3) is a stereoisomer of 7-dehydrocholesterol and is produced through the photochemical transformation of 7-dehydrocholesteol induced by high doses of UVB. L3 is enzymatically hydroxylated by CYP11A1, producing 20(OH)L3, 22(OH)L3, 20,22(OH)2L3, and 24(OH)L3. Hydroxylumisterols function as reverse agonists of
Andreia Monteiro et al.
Journal of neuroimmunology, 339, 577113-577113 (2019-11-30)
Our work consists of a pilot study to characterize circulating Th/c1, Th/c2, Th/c17, Treg and Tfh-like populations and IL-17 serum levels of relapsing-remitting (RR) MS patients treated with IFN-β, compared with healthy controls. In remission RRMS patients, we observe increased
Jinyu Zhang et al.
International immunology, 26(7), 407-415 (2014-03-22)
MiR-155 (-/-) mice are highly resistant to experimental autoimmune encephalomyelitis (EAE), while Pdcd1 (-/-) mice develop a more severe form of the disease. To determine the conflicting roles of these two molecules in the disease, we generated miR-155 (-/-) Pdcd1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service