RAB0215
Mouse HGF ELISA Kit
for cell and tissue lysates
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell lysate
sample type tissue lysate
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 400 pg/mL
standard curve range: 0.614-60 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Hgf(15234)
General description
The Mouse HGF (Hepatocyte Growth Factor) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse HGF cell lysate and tissue lysate.
Immunogen
Recombinant Mouse HGF
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Qian Wen et al.
Journal of translational medicine, 12, 114-114 (2014-06-03)
Autologous transplantation of modified mesenchymal stem cells (MSCs) is a promising candidate for the treatment of the refractory clinical disease, avascular necrosis of the femoral head (ANFH). Our previous attempts by compounding MSCs with medical fibrin glue to treat ANFH
Jesse Hudson et al.
Molecular and cellular biology, 34(19), 3689-3701 (2014-07-30)
Breast cancers are stratified into distinct subtypes, which influence therapeutic responsiveness and patient outcome. Patients with luminal breast cancers are often associated with a better prognosis relative to that with other subtypes. However, subsets of patients with luminal disease remain
Elisa Tinazzi et al.
Immunology letters, 161(2), 236-240 (2014-01-23)
CD30 and CD30 ligand (CD30L) are members of TNF-receptor and TNF superfamilies respectively. CD30(+)T cells are increased in several diseases and interaction between CD30(+) and CD30L(+)T cells leads either to cell proliferation or apoptosis. In patients with rheumatoid arthritis (RA)
Ji-Dong Chen et al.
Asian Pacific journal of cancer prevention : APJCP, 15(11), 4545-4548 (2014-06-28)
To investigate the changes of serum vascular endothelial growth factor (VEGF), soluble interleukin-2 receptor (SIL-2R) and hepatocyte growth factor (HGF) contents in patients with primary hepatocellular carcinoma (HCC) before and after percutaneous microwave coagulation therapy (PMCT) and determine their clinical
Mishaal B Patel et al.
Carcinogenesis, 35(8), 1891-1900 (2014-05-31)
Activated cancer-associated human pancreatic stellate cells (CAhPSCs, which produce the collagenous stroma of pancreatic cancer [PC]) are known to play a major role in PC progression. Apart from inducing cancer cell proliferation and migration, CAhPSCs have also been implicated in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service