Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0205

Sigma-Aldrich

Human GDNF ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type urine
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GDNF(2668)

General description

Glial cell derived neurotrophic factor (GDNF) is encoded by the gene mapped to human chromosome 5p14.1−q13.3. It belongs to the transforming growth factor β (TGF-β) superfamily. GDNF is abundantly found in gliomas.

Immunogen

Recombinant Human GDNF

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Glial cell derived neurotrophic factor (GDNF) is a neurotrophic factor involved in dopaminergic and neurodevelopment. It plays a vital role in self-renewal of stem cells. In addition, GDNF also facilitates differentiation of chained undifferentiated spermatogonia. GDNF with its receptors, activates various signaling pathways essential for maintaining cellular homeostasis. Mutated gene is associated with the development of schizophrenia. GDNF is a potential target for treating Parkinson′s disease.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association analysis of the glial cell line-derived neurotrophic factor (GDNF) gene in schizophrenia
Williams HJ, et al.
Schizophrenia Research, 97(1-3), 271-276 (2007)
Liliana Brambilla et al.
Human molecular genetics, 25(14), 3080-3095 (2016-06-12)
Considerable evidence indicates that neurodegeneration in amyotrophic lateral sclerosis (ALS) can be conditioned by a deleterious interplay between motor neurons and astrocytes. Astrocytes are the major glial component in the central nervous system (CNS) and fulfill several activities that are
The reversible effects of glial cell line--derived neurotrophic factor (GDNF) in the human brain
Ayanlaja A, et al.
Seminars in Cancer Biology, 53(3), 212-222 (2018)
Wenxing Zhao et al.
Neuroscience letters, 637, 108-113 (2016-11-27)
Inflammatory and neurotrophic factors are involved in postherpetic neuralgia (PHN), but the association of these factors in the cerebrospinal fluid (CSF) with the level of pain is poorly known. The present study aimed to examine the changes in neurotrophic and
Distribution of GFRA1-expressing spermatogonia in adult mouse testis
Grasso M, et al.
Reproduction (Cambridge, England), 143(3) (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service