MilliporeSigma
  • Changes in neurotrophic and inflammatory factors in the cerebrospinal fluid of patients with postherpetic neuralgia.
Changes in neurotrophic and inflammatory factors in the cerebrospinal fluid of patients with postherpetic neuralgia.
Neuroscience letters (2016-11-27)
Wenxing Zhao, Yong Wang, Qiwu Fang, Jianping Wu, Xinyou Gao, Hui Liu, Liu Cao, Jianxiong An
ABSTRACT
Inflammatory and neurotrophic factors are involved in postherpetic neuralgia (PHN), but the association of these factors in the cerebrospinal fluid (CSF) with the level of pain is poorly known. The present study aimed to examine the changes in neurotrophic and inflammatory factors in the CSF of patients with PHN and to study the correlation between these factors and the degree of pain. Fifty patients with PHN and 28 patients with hemifacial spasm (as controls) were recruited between May 2015 and March 2016. CSF levels of inflammatory and neurotrophic factors were measured by ELISA. Compared with controls, patients with PHN had lower CSF levels of brain-derived neurotrophic factor (BDNF), nerve growth factor (NGF), neurotrophin (NT)-3, NT-5, and P substance (all P<0.05), and higher CSF levels of interleukin (IL)-1β (P=0.050). Among patients with PHN, CSF BDNF levels were positively correlated to IL-8 (r
Sigma-Aldrich
Human NGF R  ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human GDNF ELISA Kit, for serum, plasma, cell culture supernatant and urine