Skip to Content
MilliporeSigma
All Photos(4)

Documents

R7750

Sigma-Aldrich

D-Ribose 5-phosphate disodium salt hydrate

≥98% (TLC), powder

Synonym(s):

D-Ribofuranose 5-phosphate disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9Na2O8P · xH2O
Molecular Weight:
274.07 (anhydrous basis)
Beilstein:
4774244
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥98% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

impurities

6.0-15.0% water (Karl Fischer)

color

white

solubility

water: 50 mg/mL, clear, colorless

cation traces

Na: 15.0-17.0% (dry basis)

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O

InChI

1S/C5H11O8P.2Na.H2O/c6-3-2(1-12-14(9,10)11)13-5(8)4(3)7;;;/h2-8H,1H2,(H2,9,10,11);;;1H2/q;2*+1;/p-2/t2-,3-,4-,5?;;;/m1.../s1

InChI key

HKCYZJCJVGKHNA-IREXRYMASA-L

Biochem/physiol Actions

Ribose-5-phosphate is an intermediate of the oxidative branch of the Pentose Phosphate Pathway(PPP) and an end product of the nonoxidative branch of the PPP. Ribose-5-phosphate is used in the synthesis of nucleotides and nucleic acids.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Hong et al.
Cell chemical biology, 26(4), 493-501 (2019-02-05)
Malayamycin A is an unusual bicyclic C-nucleoside, with interesting antiviral, antifungal, and anticancer bioactivity. We report here the discovery and characterization of the biosynthetic pathway to malayamycin by using genome mining of near-identical clusters both from the known producer Streptomyces
Josefin Engelhardt et al.
PloS one, 15(1), e0227201-e0227201 (2020-01-03)
The eastern Baltic cod (Gadus morhua) population has been decreasing in the Baltic Sea for at least 30 years. Condition indices of the Baltic cod have decreased, and previous studies have suggested that this might be due to overfishing, predation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service