Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R7513

Sigma-Aldrich

RPMI-1640 Medium

With sodium bicarbonate, without methionine, cystine and ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

RPMI culture medium, RPMI-1640 cell culture medium, Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

HEPES: no
sodium pyruvate: no
phenol red: yes
L-glutamine: no
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Abdullah et al.
Biomolecules & therapeutics (2021-10-21)
Targeting the cystine/glutamate exchange transporter, system xc -, is a promising anticancer strategy that induces ferroptosis, which is a distinct form of cell death mediated by iron-dependent lipid peroxidation. The concentration of L-cystine in culture medium is higher than the
Nathanael G Lintner et al.
PLoS biology, 15(3), e2001882-e2001882 (2017-03-23)
Proprotein convertase subtilisin/kexin type 9 (PCSK9) plays a key role in regulating the levels of plasma low-density lipoprotein cholesterol (LDL-C). Here, we demonstrate that the compound PF-06446846 inhibits translation of PCSK9 by inducing the ribosome to stall around codon 34
Bin Wu et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 17(4), 1878-1896 (2020-09-23)
The molecular mechanism of Alzheimer-like cognitive impairment induced by manganese (Mn) exposure has not yet been fully clarified, and there are currently no effective interventions to treat neurodegenerative lesions related to manganism. Protein phosphatase 2 A (PP2A) is a major
Ling Meng et al.
Frontiers in immunology, 12, 648913-648913 (2021-04-30)
The excessive M1 polarization of macrophages drives the occurrence and development of inflammatory diseases. The reprogramming of macrophages from M1 to M2 can be achieved by targeting metabolic events. Taurine promotes for the balance of energy metabolism and the repair
Hanlin Zhang et al.
Molecular cell, 76(1), 110-125 (2019-09-03)
Failure to make adaptive immune responses is a hallmark of aging. Reduced B cell function leads to poor vaccination efficacy and a high prevalence of infections in the elderly. Here we show that reduced autophagy is a central molecular mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service