Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R5886

SAFC

RPMI-1640 Medium

HEPES modification, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

pH

>7.2

components

L-glutamine: no
sodium pyruvate: no
HEPES: 25 mM
phenol red: yes
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.
This RPMI-1640 medium is supplemented with HEPES. HEPES is a Good′s physiological buffer that helps provide a more stable medium pH than bicarbonate buffers.

Application

RPMI-1640 Medium has been used:
  • to culture human lymphoblastoid cell line cells
  • prostate cancer cells
  • monocytes from human blood

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marga J Bouma et al.
Stem cell research, 41, 101654-101654 (2019-11-18)
Lymphoblast cells from four individuals of a family of two genetically unrelated parents and their monozygotic twins were used to generate integration-free human induced pluripotent stem cells (hiPSCs). Reprogramming factors were delivered by co-electroporation of three episomal-based plasmids expressing OCT3/4
Stephanie Puukila et al.
International journal of radiation biology, 95(8), 1072-1084 (2019-05-30)
Purpose: Diagnostic radiation is an important part of patient care in the Intensive Care Unit; however, there is little data on the acute effects of exposure to these doses. We investigated pulmonary and splenic response 30 minutes, 4 hours or 24 hours after
Serena De Lucia et al.
PloS one, 13(1), e0191084-e0191084 (2018-01-18)
Artemisinin resistance is a major threat to malaria control efforts. Resistance is characterized by an increase in the Plasmodium falciparum parasite clearance half-life following treatment with artemisinin-based combination therapies (ACTs) and an increase in the percentage of surviving parasites. The
Katrin Kleinmanns et al.
EBioMedicine, 56, 102783-102783 (2020-05-27)
The completeness of resection is a key prognostic indicator in patients with ovarian cancer, and the application of tumour-targeted fluorescence image-guided surgery (FIGS) has led to improved detection of peritoneal metastases during cytoreductive surgery. CD24 is highly expressed in ovarian
Iveta Gažová et al.
Frontiers in cell and developmental biology, 8, 498-498 (2020-07-29)
The response of the human acute myeloid leukemia cell line THP-1 to phorbol esters has been widely studied to test candidate leukemia therapies and as a model of cell cycle arrest and monocyte-macrophage differentiation. Here we have employed Cap Analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service