Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R2758

Microcentrifuge tube rack, rectangular

6 x 12 400μl tubes, white

Synonym(s):

tube holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.35

material

polypropylene

feature

6 x 12 400μl tubes

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F18840-0000

L × W × H

9 in. × 4 3/8 in. × 4 3/8 in.

color

white

Looking for similar products? Visit Product Comparison Guide

General description

Stackable racks hold 72 tubes in a 6 × 12 array.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service