Key Documents
HS120243
Microtube Rack
blue Nylon, pack of 4 racks, Holds 96 x 1.5-2 mL tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.22
material
blue Nylon
feature
Holds 96 x 1.5-2 mL tubes
packaging
pack of 4 racks
manufacturer/tradename
Heathrow Scientific HS120243
technique(s)
PCR: suitable
well diam.
11.1 mm
Features and Benefits
  • made from reinforced nylon
  • superior chemical and heat resistance
  • handles provide easy and safe transportation
  • stackable two formats available for smaller and larger requirements
  • temperature range -40 to 140C
  • autoclavable
Choose from one of the most recent versions:
Certificates of Analysis (COA)