Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R2283

Sigma-Aldrich

Pepsin Reagent, Ready to Use, Antigen Retriever

suitable for immunohistochemistry

Synonym(s):

pepsin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

biological source

Porcine stomach

Quality Level

application(s)

hematology
histology

storage temp.

2-8°C

Application

Pepsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies in immunohistochemistry (IHC).
Pepsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In immunohistochemistry (IHC), most commonly used fixatives such as formalin mask tissue antigens (cellular, membrane, and nuclear) by their intrinsic crosslinking. This masking results in poor or no staining in IHC. Pepsin digestion of FFPE tissue sections improves accessibility of antibodies to tissue antigens. Ready to use pepsin reagent, antigen retriever has been used for antigen unmasking in immunohistochemistry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Codistribution of collagen type IV and laminin in liver fibrosis of elderly cadavers: immunohistochemical marker of perisinusoidal basement membrane formation.
Mak Ki M, et al.
The Anatomical Record, 296(6)A, 953-964 (2013)
The effect of low doses of cadmium and zinc chloride on the Blood-Testis Barrier of Sprague Dawley rats.
Redondo A, et al.
arXiv (2012)
Fetal testis organ culture reproduces the dynamics of epigenetic reprogramming in rat gonocytes.
Rwigemera A, et al.
Epigenetics & Chromatin, 10(1), 19-19 (2017)
M Bekhouche et al.
Journal of materials chemistry. B, 8(36), 8422-8432 (2020-08-18)
Hydrogel-based regenerative endodontic procedures (REPs) are considered to be very promising therapeutic strategies to reconstruct the dental pulp (DP) tissue in devitalized human teeth. However, the success of the regeneration process is limited by residual bacteria that may persist in
Elizabeth Vinod et al.
Acta histochemica, 122(1), 151462-151462 (2019-11-18)
Interest in chondroprogenitors arose due to their inherent stem cell like properties, and their initial characterization was based on identification of a small percentage of CD49e positive cells in cultured chondrocytes (CC). It was further noted that when fresh chondrocytes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service