Skip to Content
MilliporeSigma
All Photos(1)

Documents

10108057001

Roche

Pepsin

lyophilized (salt-free), ~2500 units/mg protein (At 37 °C with hemoglobin as the substrate. One unit is the enzyme activity which liberates the amount of Tyr producing an increase in the absorbance of 0.001/minute at 280 nm.)

Synonym(s):

pepsin

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

pig gastric mucosa

Quality Level

form

lyophilized (salt-free)

specific activity

~2500 units/mg protein

packaging

pkg of 1 g

manufacturer/tradename

Roche

optimum pH

1.8-2.2

shipped in

wet ice

storage temp.

2-8°C

General description

Pepsin is an aspartic endoproteinase isolated from pig gastric mucosa. It is mainly used for the unspecific hydrolysis of proteins and peptides in acidic media. It also provides a limited hydrolysis of native immunoglobulins to yield biologically active fragments.

Specificity

Aspartic endopeptidase with relatively broad specificity. Preferential cleavage: hydrophobic, preferably aromatic, residues in P1 and P1′ positions.

Application

Pepsin has been used for the digestion of liver tissue obtained from rat.

Preparation Note

Working concentration: 0.05 to 0.5 mg/ml
Storage conditions (working solution): A pepsin solution is stable at 2 to 8 °C, at least for one week, at neutral pH and under germfree conditions. Frozen aliquots of the enzyme solution are expected to be more stable.

A stock solution in 10 mM HCl (5-10 mg/ml) should be stable for at least one week at 2 to 8 °C, too. Storage light protected, tightly closed.

A short (2 minutes) incubation at 40 °C has negligible effect on the activity of lyophilized pepsin or pepsin in solution.

Analysis Note

At 37 °C with hemoglobin as the substrate. One unit is the enzyme activity which liberates the amount of Tyr producing an increase in the absorbance of 0.001/minute at 280 nm.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren M Ashwood et al.
BMC biology, 21(1), 121-121 (2023-05-25)
The ShK toxin from Stichodactyla helianthus has established the therapeutic potential of sea anemone venom peptides, but many lineage-specific toxin families in Actiniarians remain uncharacterised. One such peptide family, sea anemone 8 (SA8), is present in all five sea anemone superfamilies.
Marie van Dijk et al.
Prenatal diagnosis, 42(13), 1612-1621 (2022-11-08)
Noninvasive Prenatal Diagnosis has recently been introduced for a limited number of monogenetic disorders. However, the majority of DNA diagnostics still require fetal material obtained using an invasive test. Recently, a novel technique, TRIC (Trophoblast Retrieval and Isolation from the
Ganesan Arunkumar et al.
Science advances, 8(9), eabl5621-eabl5621 (2022-03-03)
Chromosome instability is a critical event in cancer progression. Histone H3 variant CENP-A plays a fundamental role in defining centromere identity, structure, and function but is innately overexpressed in several types of solid cancers. In the cancer background, excess CENP-A
William D Kim et al.
Frontiers in genetics, 13, 1045738-1045738 (2022-11-29)
Mutations in CLN5 cause a subtype of neuronal ceroid lipofuscinosis (NCL) called CLN5 disease. The NCLs, commonly referred to as Batten disease, are a family of neurodegenerative lysosomal storage diseases that affect all ages and ethnicities globally. Previous research showed
Claus Kordes et al.
The Journal of clinical investigation, 124(12), 5503-5515 (2014-11-18)
Retinoid-storing hepatic stellate cells (HSCs) have recently been described as a liver-resident mesenchymal stem cell (MSC) population; however, it is not clear whether these cells contribute to liver regeneration or serve as a progenitor cell population with hepatobiliary characteristics. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service