Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R1153

Sigma-Aldrich

RNase B Glycoprotein Standard from bovine pancreas

≥90% (SDS-PAGE), lyophilized powder

Synonym(s):

Ribonuclease B from bovine pancreas, RNase B

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

Product Name

RNase B Glycoprotein Standard from bovine pancreas, Proteomics Grade

grade

Proteomics Grade

Quality Level

Assay

≥90% (SDS-PAGE)

form

lyophilized powder

storage temp.

−20°C

SMILES string

[nH]1cnc(c1)CC(NC(=O)CCN)C(=O)O

InChI

1S/C9H14N4O3/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6/h4-5,7H,1-3,10H2,(H,11,12)(H,13,14)(H,15,16)

InChI key

CQOVPNPJLQNMDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

RNase B is a preferred substrate with PNGase F for demonstration of N-linked deglycosylation using SDS-PAGE or MALDI-MS. The activity of PNGase F is routinely assayed using RNase B by monitoring the pronounced mobility shift in 12% gels after deglycosylation. Proteomics Grade RNase B has been used as a source of N-glycans following enzymatic digestions and subsequent purification. It has also been used as a glycoprotein standard.

Other Notes

Bovine pancreatic RNase B is a glycoprotein that contains only N-linked glycans. It is a globular protein composed of a single domain that occurs naturally as a lesser component in mixture along with ribonuclease A (RNase A), which is the non-glycosylated core form. RNase B contains a single glycosylation site at Asn34 at which from five to nine mannose residues are attached to the chitobiose core, i.e. Man5-9GlcNAc2. Due to the heterogeneity in the glycosylation at Asn34, RNase B exists as five glycosylated varients, with a molecular weight of approx. 15 kDa.

Quality

RNAse B Glycoprotein Standard has been highly purified to remove contaminating RNase A.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrated GlycoProteome Analyzer (I-GPA) for automated identification and quantitation of site-specific N-glycosylation
Park GW, et al.
Scientific Reports, 6, 21175-21175 (2016)
Cytosolic nuclease TREX1 regulates oligosaccharyltransferase activity independent of nuclease activity to suppress immune activation
Hasan M, et al.
Immunity, 43(3), 463-474 (2015)
Fabian Higel et al.
mAbs, 6(4), 894-903 (2014-05-23)
N-glycosylation is a complex post-translational modification with potential effects on the efficacy and safety of therapeutic proteins and known influence on the effector function of biopharmaceutical monoclonal antibodies (mAbs). Comprehensive characterization of N-glycosylation is therefore important in biopharmaceutical development. In
Maroof Hasan et al.
Immunity, 43(3), 463-474 (2015-09-01)
TREX1 is an endoplasmic reticulum (ER)-associated negative regulator of innate immunity. TREX1 mutations are associated with autoimmune and autoinflammatory diseases. Biallelic mutations abrogating DNase activity cause autoimmunity by allowing immunogenic self-DNA to accumulate, but it is unknown how dominant frameshift
Alex M Yoshikawa et al.
Nature communications, 12(1), 7106-7106 (2021-12-09)
Glycosylation is one of the most abundant forms of post-translational modification, and can have a profound impact on a wide range of biological processes and diseases. Unfortunately, efforts to characterize the biological function of such modifications have been greatly hampered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service