Skip to Content
MilliporeSigma
All Photos(1)

Documents

112251

Sigma-Aldrich

α₁-Acid Glycoprotein, Human Plasma

Synonym(s):

α₁-Acid Glycoprotein, Human Plasma, α₁-AG, C-AGP, Orosomucoid, α₁M-Seromucoid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

human plasma

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Native α1-acid glycoprotein from human plasma. Glycoprotein used for the study of the structure of oligosaccharide units. Consists of 40 - 45% carbohydrate. Present in human plasma at concentrations of 55 - 140 mg/100 ml. Clinically,α1-AG is an acute-phase reactant that, together with haptoglobin, indicates acute inflammation. Selectively suppresses the augmentation of NK cell activity by IFN-α and IFN-γ. Theα1-AG:α1-antitrypsin ratio is useful in diagnosis of liver disease. A sialic acid-deficientα1-AG has been observed in serum or urine of patients with Hodgkin’s disease, psoriatic arthritis, diabetes, and chronic myeloid leukemia. Sialic acid-deficientα1-AG has an affinity for vitamin B12.
Native α1-acid glycoprotein from human plasma. Glycoprotein used for the study of the structure of oligosaccharide units. Consists of 40-45% carbohydrate. Present in human plasma at concentrations of 55-140 mg/100 ml. Clinically, α1-AG is an acute phase reactant that, together with haptoglobin, indicates acute inflammation. Selectively suppresses the augmentation of NK cell activity by IFN-α and IFN-γ. The α1-AG:α1-antitrypsin ratio is useful in diagnosis of liver disease. A sialic acid-deficient α1-AG has been observed in serum or urine of patients with Hodgkin′s disease, psoriatic arthritis, diabetes, and chronic myeloid leukemia. Sialic acid-deficient α1-AG has an affinity for vitamin B12.

Warning

Toxicity: Standard Handling (A)

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for to 3 months at (-20°C).

Other Notes

Aso, H., et al. 1999. Inflammation 23, 117.
Schmid, K., et al. 1973. Biochemistry 12, 2711.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service